missing translation for 'onlineSavingsMsg'
Learn More
Di-potassium oxalate monohydrate, ExpertQ, Scharlab Product Code.: 11334274
compliance-icons

Di-potassium oxalate monohydrate, ExpertQ, Scharlab

Product Code. 11334274
500 g, 500g
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11334274

Brand: Scharlab PO03100500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6487-48-5
Quantity 500 g
Molecular Formula C2H2K2O5
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O
IUPAC Name dipotassium hydrate oxalate
Molecular Weight (g/mol) 184.23
Grade ExpertQ
compliance-icons
Product Identifier
  • di-Potassium oxalate monohydrate, reagent grade, ACS
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-potassium oxalate monohydrate, ExpertQ, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.