missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, Ultrapure
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
Specifications
Specifications
Physical Form | Powder |
Quantity | 100 g |
Formula Weight | 60,000- 90,000 |
Packaging | Plastic bottle |
Chemical Name or Material | Dextran |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
192419 | Certificate of Analysis | 20/10/2023 | J14495.22,J14495.A1 |
195107 | Certificate of Analysis | 20/10/2023 | J14495.22,J14495.A1 |
177969 | Certificate of Analysis | 27/11/2022 | J14495.22,J14495.A1 |
197585 | Certificate of Analysis | 27/11/2022 | J14495.22,J14495.A1 |
195508 | Certificate of Analysis | 27/11/2022 | J14495.22,J14495.A1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dextran, Ultrapure >
Spot an opportunity for improvement?Share a Content Correction