Learn More
Lenz Laborglasintrumente Delivery Adapters Straight with Suction Tube
Click to view available options
Standard Taper Joint No.:
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
Description
- Delivery adapters
- Straight
- With suction tube
- With NS socket and NS cone
- With drain tip
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Standard Taper Joint No. | NS 14/23 |