Learn More
Eppendorf™ Deepwell Plate Rotor for A-2-DWP Benchtop Centrifuge
Use this rotor with Eppendorf™ model A-2-DWP Benchtop Centrifuge. Eppendorf™ Deepwell Plate Rotor for A-2-DWP Benchtop Centrifuge includes two deepwell plate buckets.
Brand: Eppendorf™ 5804740009
22032.78 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
- For use with: A-2-DWP Benchtop Centrifuge
Specifications
Deepwell Plate Rotor | |
4,000 rpm | |
2 buckets |
2,250 x g | |
A-2-DWP |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
57Hours
7Minutes
20Seconds
59 -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31