Learn More
Borer Chemie™ deconex 22 LIQ-x
Universal use for automated cleaning of laboratory glassware and utensils. Without phosphates, surfactants and chlorine. deconex 22 LIQ-x is an ecological solution with an excellent performance. Acidic rinse with a deconex product recommended.
Brand: Borer Chemie™ 519200.00-K25W
This item is not returnable.
View return policy
Description
- Concentrate for universal automated cleaning
- Ecological solution without phosphates and surfactants
- Without chlorine, EDTA or NTA
- High yield, typical concentration: 5-10 mL/L
- Broad cleaning spectrum
- High performance thanks to high dispersing capacity
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Acid rinse with a deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, sequestering agents | |
Canister | |
1 x 30 kg | |
24.4 L | |
30 kg |
∼12.2 (1% Solution) | |
Concentrate | |
5°C to 25°C | |
6.4 gal. | |
66.1 lb. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- deconex 22 LIQ-x
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : potassium hydroxide,DIPOTASIUM TRIOXOSILIC