Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Brand:  Trajan Scientific and Medical™ 062446
 View more versions of this product
Product Code. 15278803
  • 518.00 NOK / 2m
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Description
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifications
Specifications
Organic and Aqueous Solvents
1/Pk.
170 μm
Thermally stable up to 380°C
Tubing
110 μm
2 m
Product Title
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing > Deactivated Fused Silica Tubing; 170μm O.D. x 110μm I.D. x 2m L; 1/pack