Learn More
Thermo Scientific™ Deactivated Glass Wool
Pack GC injection port liners with deactivated glass wool for good homogenization, volatilization of high boiling point compounds, and minimal column contamination.
Brand: Thermo Scientific™ 60180-785
879.98 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Pack your GC injection port liner with Thermo Scientific™ deactivated glass wool for good homogenization and volatilization of high boiling point compounds within the liner. The glass wool acts as a barrier to prevent non-volatile sample matrix compounds and septa particles from entering GC columns, minimizing contamination and increasing precision and reproducibility.
Specifications
Deactivated Glass Wool 10 g | |
10 g | |
Deactivated Glass Wool | |
GC Liners |