missing translation for 'onlineSavingsMsg'
Learn More

Daclatasvir dihydrochloride, MedChemExpress

MedChemExpress Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

Encompass_Preferred

821.80 NOK - 1469.75 NOK

Specifications

Chemical Name or Material Daclatasvir dihydrochloride
CAS 1009119-65-6
Color White
Purity Grade Notes Research
Molecular Formula C40H52Cl2N8O6
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16496697
Encompass_Preferred
View Documents
Medchem Express
HY-10465
5 mg
821.80 NOK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16473988
Encompass_Preferred
View Documents
Medchem Express
HY-10465
10 mg
979.83 NOK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16420347
Encompass_Preferred
View Documents
Medchem Express
HY-10465
50 mg
1311.71 NOK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16424996
Encompass_Preferred
View Documents
Medchem Express
HY-10465
100 mg
1469.75 NOK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Specifications

Specifications

Daclatasvir dihydrochloride
White
C40H52Cl2N8O6
DMSO : ≥ 56 mg/mL (68.98 mM) ∣H2O : 50 mg/mL (61.59 mM; Need ultrasonic)
811.8
95.0%
Solid
1009119-65-6
Research
BMS-790052 dihydrochlorideEBP 883 dihydrochloride
O=C(N1CCC[C@H]1C2=NC=C(C3=CC=C(C4=CC=C(C=C4)C5=CN=C(N5)[C@@H]6CCCN6C([C@H](C(C)C)NC(OC)=O)=O)C=C3)N2)[C@H](C(C)C)NC(OC)=O.[H]Cl.[H]Cl
811.8
Research
COVID-19-anti-virus
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Daclatasvir dihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.