missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Prolinamide, 98%, Thermo Scientific Chemicals
865.80 NOK - 2876.40 NOK
Chemical Identifiers
CAS | 62937-45-5 |
---|---|
Molecular Formula | C5H10N2O |
Molecular Weight (g/mol) | 114.15 |
MDL Number | MFCD00153457 |
InChI Key | VLJNHYLEOZPXFW-UHFFFAOYNA-N |
Synonym | d-prolinamide, r-prolinamide, h-d-pro-nh2, r-pyrrolidine-2-carboxamide, 2r-pyrrolidine-2-carboxamide, d-proline amide, h-d-pro-nh, d-pro-nh2, 2r-2-carbamoylpyrrolidine, 2-pyrrolidinecarboxamide, 2r |
PubChem CID | 447554 |
IUPAC Name | (2R)-pyrrolidine-2-carboxamide |
SMILES | NC(=O)C1CCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463997
|
Thermo Scientific Alfa Aesar
H30901.03 |
1 g |
865.80 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473997
|
Thermo Scientific Alfa Aesar
H30901.06 |
5 g |
2876.40 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
62937-45-5 | |
114.15 | |
VLJNHYLEOZPXFW-UHFFFAOYNA-N | |
447554 | |
NC(=O)C1CCCN1 |
C5H10N2O | |
MFCD00153457 | |
d-prolinamide, r-prolinamide, h-d-pro-nh2, r-pyrrolidine-2-carboxamide, 2r-pyrrolidine-2-carboxamide, d-proline amide, h-d-pro-nh, d-pro-nh2, 2r-2-carbamoylpyrrolidine, 2-pyrrolidinecarboxamide, 2r | |
(2R)-pyrrolidine-2-carboxamide |
Specifications
62937-45-5 | |
C5H10N2O | |
d-prolinamide, r-prolinamide, h-d-pro-nh2, r-pyrrolidine-2-carboxamide, 2r-pyrrolidine-2-carboxamide, d-proline amide, h-d-pro-nh, d-pro-nh2, 2r-2-carbamoylpyrrolidine, 2-pyrrolidinecarboxamide, 2r | |
VLJNHYLEOZPXFW-UHFFFAOYNA-N | |
NC(=O)C1CCCN1 | |
114.15 | |
114.15 | |
D-(-)-Prolinamide |
100°C to 103°C | |
MFCD00153457 | |
Soluble in water. | |
+107° (c=2 in Ethanol) | |
(2R)-pyrrolidine-2-carboxamide | |
447554 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only