missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D(+)-Mannose, 99+% Product Code.: 11926901

Thermo Scientific Chemicals D(+)-Mannose, 99+%

Product Code. 11926901 Shop All Thermo Scientific Chemicals Products
2.5 kg, Plastic drum, 2.50kg
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
2.50kg
25g
This item is not returnable. View return policy

Product Code. 11926901

Brand: Thermo Scientific Chemicals 150600025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 133.0°C
Color White
Loss on Drying 0.45% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Odor Odorless
Packaging Plastic drum
Quantity 2.5 kg
Beilstein 31284
Merck Index 155812
Specific Rotation Condition + 14.00 (20.00°C 589nm,c=10,H2O)
Specific Rotation + 14.00
Solubility Information Solubility in water: 2480g/L (17°C)
Formula Weight 180.16
Percent Purity 99+%
Physical Form Crystals or Powder
Chemical Name or Material D(+)-Mannose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0471733 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code150600250,150601000,150600010,150600025
Lot NumberA0471734 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code150600250,150601000,150600010,150600025
Lot NumberA0471735 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code150600250,150601000,150600010,150600025
Lot NumberA0438417 Certificate TypeCertificate of Analysis Date07/02/2024 Product Code150600250,150600010,150601000,150600025
Lot NumberA0438416 Certificate TypeCertificate of Analysis Date05/02/2024 Product Code150600250,150600010,150601000,150600025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D(+)-Mannose, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.