Learn More
MedChemExpress D-JNKI-1
MedChemExpress D-JNKI-1 (AM-111) is a highly potent and cell-permeable peptide inhibitor of JNK.
Brand: MedChemExpress HY-P0069
Specifications
95.04% | |
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month | |
Reference compound | |
Room temperature | |
AM-111∣ XG-102 |
1445179-97-4 | |
3822.44 g/mol | |
5 mg | |
DMSO : ≥ 100 mg/mL (26.16 mM) ∣H2O : ≥ 50 mg/mL (13.08 mM) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.