missing translation for 'onlineSavingsMsg'
Learn More
D(+)-Glucose Anhydrous, ACS Reagent Grade, Scharlab Product Code.: 11384144

D(+)-Glucose Anhydrous, ACS Reagent Grade, Scharlab

Product Code. 11384144
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11384144

Brand: Scharlab GL01271000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, in biochemistry, for pharmaceutical use, in the food industry and in the pharmaceutical industry.

  • ACS Reagent Grade
  • Synonyms: Dextrose
  • Molecular Formula: C6H1206
  • Molar Mass: 180.16g/mol
  • CAS: 50-99-7
  • EIN ECS No.: 200-075-1
  • Starch: Passes test

Specifications

Acidity Acidity max: 0.002mEq/g
CAS 50-99-7
Melting Point ∼146°C
Ignition Residue 0.01% max
Loss on Drying 0.2% max (105°C)
Ignition Point ∼500°C
Molecular Formula C6H12O6
Quantity 1 kg
Specific Rotation 52.5–53.0° ([a]25°/D, c=10, water)
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
Solubility Information In water: ∼470g/L at 20°C
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILES C(C(C(C(C(C=O)O)O)O)O)O
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.156
PubChem CID 107526
ChEBI CHEBI:42758
Formula Weight 180.16g/mol
Grade ACS Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D(+)-Glucose Anhydrous, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.