Learn More
IBA Lifesciences Desthiobiotin
GREENER_CHOICE
Desthiobiotin powder for preparation of Strep-Tactin™ elution buffer
Brand:  IBA Lifesciences 2-1000-002
 View more versions of this product
Product Code. 12753064
  • 2500.00 NOK / Each
Description
Desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
Desthiobiotin powder can be used for preparation of Strep-Tactin™ elution buffer, Buffer E.
Please note that desthiobiotin is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Specifications
1 g
For Elution of Strep-Tag II Proteins From Strep-Tactin
Desthiobiotin
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
IBA Lifesciences Desthiobiotin > Quantity: 1g
For Research Use Only.