Learn More
Thermo Scientific Chemicals D-allo-Isoleucine, 97%
1726.98 NOK - 2901.32 NOK
Chemical Identifiers
CAS 1509-35-9
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00066445
InChI Key AGPKZVBTJJNPAG-CRCLSJGQSA-N
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
PubChem CID 94206
ChEBI CHEBI:20899
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](N)C(O)=O
View More Specs
Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11985341
View Documents Promotion Details
Thermo Scientific Chemicals
357435000
500 mg Glass Bottle
1726.98 NOK
500mg
11975341
View Documents Promotion Details
Thermo Scientific Chemicals
357430010
1 g Glass bottle
2901.32 NOK
1g
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
1509-35-9
131.18
AGPKZVBTJJNPAG-CRCLSJGQSA-N
94206
(2R,3S)-2-amino-3-methylpentanoic acid
C6H13NO2
MFCD00066445
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
CHEBI:20899
CC[C@H](C)[C@@H](N)C(O)=O
Specifications
1509-35-9
White
97%
MFCD00066445
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
CC[C@H](C)[C@@H](N)C(O)=O
131.18
CHEBI:20899
97%
D-allo-Isoleucine
285.0°C
Authentic
C6H13NO2
− 35.00
AGPKZVBTJJNPAG-CRCLSJGQSA-N
(2R,3S)-2-amino-3-methylpentanoic acid
94206
131.18
Crystalline Powder
Videos
Safety and Handling
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 216-143-9
Product Identifier
  • D-Alloisoleucine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.