Learn More
Cytokeratin 17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18502-1-AP-20UL
 View more versions of this product
Product Code. 16898514
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.
Specifications
Specifications
Cytokeratin 17
Polyclonal
Unconjugated
KRT17
39.1, CK 17, Cytokeratin 17, K17, keratin 17, KRT17, PC, PC2, PCHC1
Rabbit
Antigen Affinity Chromatography
RUO
16667, 287702, 3872
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04695, Q6IFU8, Q9QWL7
Krt17
Cytokeratin 17 Fusion Protein Ag13351
20 μL
Primary
Rat, Mouse, Human