missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Mouse anti-Human, Rat, Clone: 1E10G2, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | Cytokeratin 13 |
---|---|
Clone | 1E10G2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848795
|
Proteintech
66684-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16838795
|
Proteintech
66684-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 13 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This type I cytokeratin is paired with keratin 4 and expressed in the suprabasal layers of non-cornified stratified epithelia. Mutations in this gene and keratin 4 have been associated with the autosomal domit disorder White Sponge Nevus. The type I cytokeratins are clustered in a region of chromosome 17q21.2. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been described.Specifications
Cytokeratin 13 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CK 13, CK13, Cytokeratin 13, K13, keratin 13, KRT13 | |
Krt13 | |
IgG1 | |
Protein G | |
Antibody |
1E10G2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P13646, Q6IFV4 | |
287699, 3860 | |
Cytokeratin 13 Fusion Protein Ag0217 | |
Primary | |
-20°C | |
KRT13 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytokeratin 13 Mouse anti-Human, Rat, Clone: 1E10G2, Proteintech