Learn More
CYP2E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19937-1-AP-20UL
 View more versions of this product
Product Code. 16859414
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
CYP2E1 can be detected as 50-57 kDa (full-length) and ∽40 kDa (N-terminal signal peptide cleaved). The antibody is specific to CYP2E1.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
Specifications
Specifications
CYP2E1
Polyclonal
Unconjugated
CYP2E1
4 nitrophenol 2 hydroxylase, CPE1, CYP2E, CYP2E1, CYP-2E1, CYPIIE1, Cytochrome P450 2E1, Cytochrome P450 J, P450 J, P450C2E
Rabbit
Antigen Affinity Chromatography
RUO
13106, 1571, 25086
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.61 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05181, P05182, Q05421
Cyp2e1
Peptide
20 μL
Primary
Rat, Human, Mouse