missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19937-1-AP-150UL
This item is not returnable.
View return policy
Description
CYP2E1 can be detected as 50-57 kDa (full-length) and ∽40 kDa (N-terminal signal peptide cleaved). The antibody is specific to CYP2E1.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.Specifications
CYP2E1 | |
Polyclonal | |
Unconjugated | |
CYP2E1 | |
4 nitrophenol 2 hydroxylase, CPE1, CYP2E, CYP2E1, CYP-2E1, CYPIIE1, Cytochrome P450 2E1, Cytochrome P450 J, P450 J, P450C2E | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13106, 1571, 25086 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05181, P05182, Q05421 | |
Cyp2e1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction