Learn More
Hellma™ Cylindrical cell far UV quartz 2mm light path 560μL Hellma
Brand:  Hellma™ 120-000-2-40
Product Code. 12791695
  • ONLINE EXCLUSIVE PRICE
    4370.00 NOK
    3088.48 NOK / Each
    Save 1281.52 NOK
    29% Off
Only null left
Estimated Shipment: 16-12-2025
Specifications
Quartz
2 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hellma™ Cylindrical cell far UV quartz 2mm light path 560μL Hellma >