Learn More
Hellma™ Cylindrical cell far UV quartz 0.5mm light path 210μL Hellma
Brand:  Hellma™ 121-0.50-40
Product Code. 12741705
  • ONLINE EXCLUSIVE PRICE
    8235.00 NOK
    5697.31 NOK / Each
    Save 2537.69 NOK
    30% Off
Only null left
Estimated Shipment: 02-01-2026
Specifications
Quartz
0.5 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hellma™ Cylindrical cell far UV quartz 0.5mm light path 210μL Hellma >