Learn More
Hellma™ Cylindrical cell far UV quartz 0.2mm light path 170μL Hellma
Brand:  Hellma™ 121-0.20-40
Product Code. 12731705
  • ONLINE EXCLUSIVE PRICE
    8235.00 NOK
    5697.31 NOK / Each
    Save 2537.69 NOK
    30% Off
Only null left
Estimated Shipment: 02-01-2026
Specifications
Quartz
0.2 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hellma™ Cylindrical cell far UV quartz 0.2mm light path 170μL Hellma >