Learn More
Saint-Gobain Cylinder measuring spouted PFA graduated 30mL Chemware
Brand: Saint-Gobain D1069601
This item is not returnable.
View return policy
Description
Specifications
30 mL |
Product Content Correction
Product Title
Saint-Gobain Cylinder measuring spouted PFA graduated 30mL Chemware >