missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Monohydrate, MP Biomedicals

Product Code. 11442142 Shop All MP Biomedicals Products
25 g
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6055-19-2
Molecular Formula C7H17Cl2N2O3P
Molecular Weight (g/mol) 279.10
MDL Number MFCD00149395
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
PubChem CID 22420
ChEBI CHEBI:4026
IUPAC Name 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1

Specifications

Assay Percent Range >98%
Quantity 25 g
Formula Weight 279.1
Chemical Name or Material Cyclophosphamide Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclophosphamide Monohydrate, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.