missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide, 99.5%, MP Biomedicals™

Product Code. 11412142
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 6055-19-2
Color White
Assay Percent Range 0.995
Molecular Formula C7H17Cl2N2O3P
MDL Number MFCD00149395
Quantity 1 g
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
IUPAC Name 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
Molecular Weight (g/mol) 279.10
PubChem CID 22420
ChEBI CHEBI:4026
Formula Weight 279.1
Physical Form Crystalline Powder
Chemical Name or Material Cyclophosphamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'