missing translation for 'onlineSavingsMsg'
Learn More
Cyclopentanecarboxaldehyde, 98%, stab. Product Code.: 15412806

Cyclopentanecarboxaldehyde, 98%, stab.

Product Code. 15412806
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15412806

Brand: Thermo Scientific Alfa Aesar H56609.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 872-53-7 | C6H10O | 98.145 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00060798
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehydeShow MoreShow Less
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O

Specifications

Density 0.919
Boiling Point 140°C to 141°C
Flash Point 28°C (82°F)
Refractive Index 1.443
Quantity 1 g
UN Number UN1989
Solubility Information Soluble in water and ethanol
Formula Weight 98.14
Percent Purity 98%
Chemical Name or Material Cyclopentanecarboxaldehyde, stabilized with 25ppm 1,4-benzoquinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ16L044 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeH56609.03,H56609.06
Lot NumberW19J009 Certificate TypeCertificate of Analysis Date04/10/2023 Product CodeH56609.03,H56609.06
Lot NumberN11C006 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56609.03,H56609.06
Lot NumberQ14G016 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56609.03,H56609.06
Lot NumberU01E055 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56609.03,H56609.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentanecarboxaldehyde, 98%, stab. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.