missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | Cyclin E1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858713
|
Proteintech
11554-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848713
|
Proteintech
11554-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are expressed only in tumors but not in the normal tissues, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.Specifications
Cyclin E1 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P24864, Q61457 | |
12447, 898 | |
Cyclin E1 Fusion Protein Ag2110 | |
Primary | |
-20°C | |
CCNE1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCNE, CCNE1, cyclin E1, G1/S specific cyclin E1 | |
CCNE1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title