missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Rabbit anti-Human, Clone: CCND1/3370R, Novus Biologicals™
Rabbit Monoclonal Antibody
2685.00 NOK
Specifications
Antigen | Cyclin D1 |
---|---|
Clone | CCND1/3370R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Cyclin D1 Monoclonal specifically detects Cyclin D1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cyclin D1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
10 mM PBS with 0.05% BSA | |
595 | |
IgG | |
Protein A or G purified |
CCND1/3370R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
A synthetic peptide corresponding to residues within aa 270-295 of Cyclin D1 | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title