missing translation for 'onlineSavingsMsg'
Learn More

Cyclin B2 Rabbit anti-Human, Mouse, Rat, Clone: 1L9B9, Novus Biologicals™

Rabbit Monoclonal Antibody

1815.00 NOK - 4580.00 NOK

Specifications

Antigen Cyclin B2
Clone 1L9B9
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18326996
View Documents
Bio-Techne
NBP3-16663-20UL
20 ÎĽg
1815.00 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
18356542
View Documents
Bio-Techne
NBP3-16663-100UL
100 ÎĽg
4580.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Cyclin B2
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cyclin B2 (O95067).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
1L9B9
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
PBS, 0.05% BSA, 50% glycerol, pH7.3
9133
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclin B2 Rabbit anti-Human, Mouse, Rat, Clone: 1L9B9, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.