missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Antibody (CCNA2/2333), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08789AF647
This item is not returnable.
View return policy
Description
Cyclin A2 Monoclonal antibody specifically detects Cyclin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cyclin A2 | |
Monoclonal | |
Alexa Fluor 647 | |
CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
Full length recombinant Cyclin A2 protein (Uniprot: P20248) | |
100 μg | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
890 | |
Store at 4°C in the dark. | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
CCNA2/2333 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction