missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL10/IP10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CXCL10/IP10 |
---|---|
Concentration | 1053 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16807663
|
Proteintech
10937-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16897653
|
Proteintech
10937-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CXCL10/IP10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL10/IP10. This antibody recognizes human antigen. The CXCL10/IP10 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
CXCL10/IP10 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |
1053 μg/mL | |
Polyclonal | |
Rabbit | |
3627 | |
Primary |