Learn More
Ambion™ Custom RNA, Stealth Seq
Custom RNA, Stealth Seq
Brand: Ambion™ 100001867
7711.70 NOK valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.Specifications
Room temperature | |
Stealth RNAI In-vivo, 500nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation | |
Stealth RNAI In-vivo | |
500 nmol |