Learn More
Ambion™ Custom RNA
Custom RNA
Brand: Ambion™ 100001868
37.83 NOK valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism. For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.Specifications
Room temperature | |
RNA In-vivo, 20nmol, Use This Tool for Fast and Easy Online Ordering of Bench-Tested Validated Stealth RNAi, siRNA, Ready-To-Order Stealth Select RNAi siRNA, Bench-Tested | |
RNA In-vivo | |
20 nmol |