Learn More
Ambion™ Custom RNA
Custom RNA
Brand: Ambion™ 100002299
35.16 NOK valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism. For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.Specifications
Block-iT™ RNAi Express, 100nmol | |
RNA DSL In-vivo | |
100 nmol |