Learn More
Custom DNA Oligo, 50N, Cartridge, Liquid
Custom DNA Oligo, 50N, Cartridge, Liquid
Brand:  Invitrogen A15608
178.00 NOK valid until 2025-06-27
Use promo code "24112" to get your promotional price.
Product Code. 15460310
  • 267.00 NOK / 50nmol
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cartridge
DNA Oligo, 50 nmol, 50N, Liquid, Cartridge
Custom DNA Oligo
50 nmol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Custom DNA Oligo, 50N, Cartridge, Liquid > 1 each