Learn More
Schneider Gerd™ Curved Dissecting Needle
Brand:  Schneider Gerd™ 652
Product Code. 10480924
  • 30.00 NOK / Each
Only null left
Estimated Shipment: 25-07-2025
Specifications
Dissecting Curved
150 x 1 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Schneider Gerd™ Curved Dissecting Needle > Size: 150 x 1mm