Learn More
Curdione, MedChemExpress
MedChemExpress Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.
Brand: Medchem Express HY-N0353
This item is not returnable.
View return policy
Description
Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.Specifications
Curdione | |
White | |
C15H24O2 | |
(+)-Curdione | |
O=C1C[C@@H](C(C)C)C(C/C(C)=C/CC[C@@H]1C)=O | |
236.35 | |
Research |
13657-68-6 | |
Research | |
10 mg | |
DMSO : ≥ 100 mg/mL (423.10 mM) | |
236.35 | |
98.61% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.