Learn More
Cupri-Tartaric Solution 1 According to European Pharmacopoeia (EP), Reagecon

High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications including use as a reagent in the performance of tests outlined in several EP monographs.
Brand: Reagecon 1023300-A
Description
- European Pharmacopoeia Reagent, Solution
- High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications.
- Applications include use as a reagent in the performance of tests outlined in several EP monographs.
- Accurate, stable and reproducible.
- Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
HDPE Bottle GL45 | |
500 mL | |
Liquid | |
EP Testing |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper sulphate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1233A24L1 | Certificate of Analysis | 14/12/2024 | 1023300-A |
1233A24K1 | Certificate of Analysis | 26/11/2024 | 1023300-A |
1233A24J1 | Certificate of Analysis | 15/10/2024 | 1023300-A |
1233A24F1 | Certificate of Analysis | 08/09/2024 | 1023300-A |
1233A24A1 | Certificate of Analysis | 21/05/2024 | 1023300-A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.