Learn More
3M™ CUNO™ Metal Screen Filter Cartridge
3M™ CUNO™ Metal Screen Filter Cartridges are made from stainless steel wire cloth for controlled particle removal efficiency.
Brand:  3M™ 7100144432
Product Code. 17302090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Strong mesh construction withstands high pressure differentials across the cartridge for filtration of high viscosity fluids
  • Elements can be cleaned chemically or ultrasonically and reused
  • Ideal for heavy viscosity liquids requiring high differential pressures to achieve specified flow rates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSL10B6A1T0100 SP GSK A0006306
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ CUNO™ Metal Screen Filter Cartridge > Model: MSL10B6A1T0100 SP GSK A0006306