Learn More
Thermo Scientific™ Culti-Loops™ Atopobium vaginae ATCC™ BAA-55™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4609450
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Product Code. 12925963
  • 2080.00 NOK / Pack of 5
Estimated Shipment: 26-07-2024
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
10124TM*
Aspergillus oryzae
Single Organism
Qualitative QC
5 Loops/Pk.
Culti-Loops are CE Marked.
Culti-Loops
Anaerobes
5 Loops/Pack
CCUG 38953 [CIP 106431, DSM 15829]
Special Offers
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.