missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL7 Mouse anti-Human, Unconjugated, Clone: 2E3G9, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CUL7 |
---|---|
Clone | 2E3G9 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16809425
|
Proteintech
67034-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16899415
|
Proteintech
67034-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CUL7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL7. This antibody recognizes human, mouse, rat antigen. The CUL7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CUL7 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
9820 | |
Primary |
2E3G9 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title