Learn More
CUL2 Mouse anti-Human, Unconjugated, Clone: 1A6G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67175-1-IG-150UL
This item is not returnable.
View return policy
Description
The CUL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL2. This antibody recognizes human, rat antigen. The CUL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CUL2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot, Immunohistochemistry, ELISA | |
1A6G9 | |
Unconjugated | |
150 μL | |
8453 | |
IgG1 |