Learn More
CUL2 Mouse anti-Human, Unconjugated, Clone: 1A6G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67175-1-IG-20UL
 View more versions of this product
Product Code. 16849665
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The CUL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL2. This antibody recognizes human, rat antigen. The CUL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CUL2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunohistochemistry, ELISA
1A6G9
Unconjugated
20 μL
8453
IgG1