Learn More
CleanSpace™ Technology CST Neck Support
CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
Brand: CleanSpace™ Technology CST1020
This item is not returnable.
View return policy
Description
- CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
- Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
- Made of light weight polypropylene plastic.
- Clips directly into the Power Unit and is easily changed.
- A CleanSpace CST Neck Support is NOT certified intrinsically safe.
Specifications
Black | |
Polypropylene | |
Medium |
Reusable | |
Neck Support |
Product Content Correction
Product Title
CleanSpace™ Technology CST Neck Support > Size: Medium