Learn More
Thermo Scientific™ Cryoware Permanent Markers and Pens
GREENER_CHOICE
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Brand:  Thermo Scientific™ 343850
1206.00 NOK valid until 2025-07-04
Use promo code "24108" to get your promotional price.
 View more versions of this product
Product Code. 10584561
  • 1775.00 NOK / Pack of 10
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Only null left
In Stock
Description
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
Features:
  • Nunc™ and Nalgene™ cryoware markers come in a variety of colors for permanent handwritten identification of samples
  • Use these markers by hand, or for higher throughput use them in the ID Scribe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Permanent Marking Pen
Cryo Pen Permanent Marker
1
Black
10
1 Pk.
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1407528 Certificate TypeCertificate of Compliance Date11/06/2025 Catalog Number343850
Lot Number1394741 Certificate TypeCertificate of Compliance Date11/06/2025 Catalog Number343850
Lot Number1398217 Certificate TypeCertificate of Compliance Date11/06/2025 Catalog Number343850
Lot Number1425033 Certificate TypeCertificate of Compliance Date11/06/2025 Catalog Number343850
Lot Number1427590 Certificate TypeCertificate of Compliance Date11/06/2025 Catalog Number343850
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Cryoware Permanent Markers and Pens > Nunc Cryo Pen Permanent Marker, Black