Learn More
STEMCELL Technologies CryoStor™ CS2
Animal component-free, defined cryopreservation medium with 2% DMSO
Brand:  STEMCELL Technologies 07932
Encompass_Preferred
Product Code. 17119921
  • 6873.57 NOK / 100mL
Description
  • CryoStor™ CS2 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 2% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS2 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS2 is cGMP-manufactured with USP grade components
Specifications
Specifications
Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells
Cryopreservation
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies CryoStor™ CS2