Learn More
Corning™ Cryogenic Vial Grippers
Enables to easily sort or move vials maintaining sterility. Corning™ Cryogenic Vial Grippers feature unique design to grasp internal or external-thread cryogenic vials.
Marca: Corning™ 432136
617.50 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
DescripciĂłn
Cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials. Grippers enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen. 5 per case.
Especificaciones
5/Pk. | |
Vial Grippers | |
Sorting and moving cryogenic vials | |
15.2 cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promociones
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.