Learn More
Fisherbrand™ Cryogenic Dots in Laser Sheets
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
Brand: Fisherbrand™ 11010207
629.48 NOK valid until 2025-01-31
Use promo code "24160" to get your promotional price.
This item is not returnable.
View return policy
Description
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.Specifications
Assorted | |
Non-sterile | |
0.37 in. |
Dot label laser sheet | |
0.5 to 1.5mL tubes | |
9.55 mm |