Learn More
Greiner Bio-One Cryo.s Cryogenic Storage Tube with Datamatrix Code
Laser-written ECC200 Datamatrix Coding Service for cryogenic storage tubes.
Brand: Greiner Bio-One 122263-2DG
This item is not returnable.
View return policy
Description
- Strong resistance to mechanical and chemical damage
- High data capacity
- Sequential sample IDs or consistent characters
- Reed-Solomon error correction for high redundancy of encrypted information
- Each Datamatrix is pro
Specifications
Cryogenic Storage Tube | |
Natural | |
Internal | |
Screw Cap |
Datamatrix Code: 14 x 14, Sequence: G200000001, G200000002 | |
2 mL | |
Writing area |
Product Content Correction
Product Title
Greiner Bio-One Cryo.s Cryogenic Storage Tube with Datamatrix Code > 2mL; Datamatrix Code: 14 x 14, Sequence: G200000001, G200000002