missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRLF1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00009244-T01.100uL
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
CRLF1 293T Cell Transient Overexpression Lysate(Denatured) | |
46.53 | |
pCMV-CRLF1 full-length | |
100 μL | |
Human |
Western Blot | |
NM_004750.2 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CRLF1 antibody (H00009244-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |