missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREBL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18007-1-AP-150UL
This item is not returnable.
View return policy
Description
cAMP response element-binding protein-like-2 was identified in a search to find genes in a commonly deleted region on chromosome 12p13 flanked by ETV6 and CDKN1B genes, frequently associated with hematopoietic maligcies, as well as breast, non-small-cell lung and ovarian cancers. CREBL2 shares a 41% identity with CRE-binding protein over a 48-base long region which encodes the bZip domain of CREB. The bZip domain consists of about 30 amino acids rich in basic residues involved in DNA binding, followed by a leucine zipper motif involved in protein dimerization. This suggests that CREBL2 encodes a protein with DNA binding capabilities. The occurrence of CREBL2 deletion in maligcy suggests that CREBL2 may act as a tumor suppressor gene.Specifications
CREBL2 | |
Polyclonal | |
Unconjugated | |
CREBL2 | |
CREBL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1389, 232430, 362453 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60519, Q32M00, Q5BJU6 | |
Crebl2 | |
CREBL2 Fusion Protein Ag12504 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction